8-Methyl-3-nitroimidazo[1,2-a]pyridine

Lieferant: Apollo Scientific

OR450071-500MG
APOSOR450071-500MGEA 782 EUR
APOSOR450071-500MG
8-Methyl-3-nitroimidazo[1,2-a]pyridine
8-Methyl-3-nitroimidazo[1,2-a]pyridine
Formel: C₈H₇N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06496198
CAS-Nummer: 67625-32-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR