Ethyl 6-chloro-3-nitroimidazo[1,2-a]pyridine-2-carboxylate
Lieferant: Apollo Scientific
OR450072-500MG
APOSOR450072-500MGEA
782
EUR
APOSOR450072-500MG
Ethyl 6-chloro-3-nitroimidazo[1,2-a]pyridine-2-carboxylate
Ethyl 6-chloro-3-nitroimidazo[1,2-a]pyridine-2-carboxylate
Formel:
C₁₀H₈ClN₃O₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD07021399 CAS-Nummer: 67625-30-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...