4-Thioxo-1,3-thiazolidin-2-one

Lieferant: Apollo Scientific

OR450087-250MG
APOSOR450087-250MGEA 132 EUR
APOSOR450087-250MG
4-Thioxo-1,3-thiazolidin-2-one
4-Thioxo-1,3-thiazolidin-2-one
Formel: C₃H₃NOS₂
Molecular Weight: 133,192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02167269
CAS-Nummer: 4303-29-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR