4-Chlorothiazol-2-amine

Lieferant: Apollo Scientific

OR450103-250MG OR450103-100MG
APOSOR450103-250MGEA 84.1 EUR
APOSOR450103-250MG APOSOR450103-100MG
4-Chlorothiazol-2-amine
4-Chlorothiazol-2-amine
Formel: C₃H₃ClN₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD09999189
CAS-Nummer: 52107-46-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR