5-Iodothiazol-2-amine

Lieferant: Apollo Scientific

OR450111-1G
APOSOR450111-1GEA 574 EUR
APOSOR450111-1G
5-Iodothiazol-2-amine
5-Iodothiazol-2-amine
Formel: C₃H₃IN₂S
Molecular Weight: 226,039 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21728666
CAS-Nummer: 41731-32-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR