3-Amino-1-(3-bromophenyl)thiourea
Lieferant: Apollo Scientific
OR450118-250MG
APOSOR450118-250MGEA
87
EUR
APOSOR450118-250MG
3-Amino-1-(3-bromophenyl)thiourea
3-Amino-1-(3-bromophenyl)thiourea
Formel:
C₇H₈BrN₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041285 CAS-Nummer: 116567-17-0 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...