5-Anilino-1,2,3,4-thiatriazole
Lieferant: Apollo Scientific
|
Achtung
|
OR450120-250MG
APOSOR450120-250MGEA
50.9
EUR
APOSOR450120-250MG
5-Anilino-1,2,3,4-thiatriazole
5-Anilino-1,2,3,4-thiatriazole
Formel:
C₇H₆N₄S Schmelzpunkt: 145…147 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011564 CAS-Nummer: 13078-30-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...