N-(2-Bromophenyl)-1,2,3,4-thiatriazol-5-amine

Lieferant: Apollo Scientific

OR450121-250MG
APOSOR450121-250MGEA 139 EUR
APOSOR450121-250MG
N-(2-Bromophenyl)-1,2,3,4-thiatriazol-5-amine
N-(2-Bromophenyl)-1,2,3,4-thiatriazol-5-amine
Formel: C₇H₅BrN₄S
Molecular Weight: 257,11 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD32264086
CAS-Nummer: 1903963-79-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR