N-(3-Bromophenyl)-1,2,3,4-thiatriazol-5-amine

Lieferant: Apollo Scientific

OR450122-250MG
APOSOR450122-250MGEA 139 EUR
APOSOR450122-250MG
N-(3-Bromophenyl)-1,2,3,4-thiatriazol-5-amine
N-(3-Bromophenyl)-1,2,3,4-thiatriazol-5-amine
Formel: C₇H₅BrN₄S
Molecular Weight: 257,11 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD32264087
CAS-Nummer: 1904016-66-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR