N-(4-Bromophenyl)-1,2,3,4-thiatriazol-5-amine

Lieferant: Apollo Scientific

OR450123-250MG
APOSOR450123-250MGEA 139 EUR
APOSOR450123-250MG
N-(4-Bromophenyl)-1,2,3,4-thiatriazol-5-amine
N-(4-Bromophenyl)-1,2,3,4-thiatriazol-5-amine
Formel: C₇H₅BrN₄S
Molecular Weight: 257,11 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD32264088
CAS-Nummer: 68371-97-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR