3-Methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-6-sulphonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
OR45030-1G
OR45030-250MG
APOSOR45030-1GEA
434
EUR
APOSOR45030-1G
APOSOR45030-250MG
3-Methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-6-sulphonyl chloride
3-Methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-6-sulphonyl chloride
Formel:
C₈H₆ClNO₄S Schmelzpunkt: 137…139 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03033702 CAS-Nummer: 62522-63-8 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...