2-Iodo-4-methoxybenzoic acid

Lieferant: Apollo Scientific

OR45099-1G
APOSOR45099-1GEA 0 EUR
APOSOR45099-1G
2-Iodo-4-methoxybenzoic acid
2-Iodo-4-methoxybenzoic acid
Formel: C₈H₇IO₃
Schmelzpunkt: 170…176 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09909619
CAS-Nummer: 54435-09-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR