3-Amino-3-methylazetidine dihydrochloride

Lieferant: Apollo Scientific

OR451000-1G
APOSOR451000-1GEA 1160 EUR
APOSOR451000-1G
3-Amino-3-methylazetidine dihydrochloride
3-Amino-3-methylazetidine dihydrochloride
Formel: C₄H₁₀N₂·2ClH
Molecular Weight: 159,057 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09264377
CAS-Nummer: 124668-47-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR