N-(3-Azetidinyl)piperidine dihydrochloride

Lieferant: Apollo Scientific

OR451002-1G
APOSOR451002-1GEA 1160 EUR
APOSOR451002-1G
N-(3-Azetidinyl)piperidine dihydrochloride
N-(3-Azetidinyl)piperidine dihydrochloride
Formel: C₈H₁₆N₂·2ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD11617822
CAS-Nummer: 178312-57-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR