1-(3-Azetidinyl)-pyrrolidine dihydrochloride

Lieferant: Apollo Scientific

OR451003-1G
APOSOR451003-1GEA 1160 EUR
APOSOR451003-1G
1-(3-Azetidinyl)-pyrrolidine dihydrochloride
1-(3-Azetidinyl)-pyrrolidine dihydrochloride
Formel: C₇H₁₄N₂·2ClH
Molecular Weight: 199,121 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11617821
CAS-Nummer: 1024589-68-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR