1-(3-Methyl-3-azetidinyl)-pyrrolidine dihydrochloride

Lieferant: Apollo Scientific

OR451004-1G
APOSOR451004-1GEA 0 EUR
APOSOR451004-1G
1-(3-Methyl-3-azetidinyl)-pyrrolidine dihydrochloride
1-(3-Methyl-3-azetidinyl)-pyrrolidine dihydrochloride
Formel: C₈H₁₆N₂·2ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD28975193
CAS-Nummer: 1152111-80-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR