tert-Butyl azetidine-2-carboxylate hydrochloride

Lieferant: Apollo Scientific

OR451006-1G
APOSOR451006-1GEA 1250 EUR
APOSOR451006-1G
tert-Butyl azetidine-2-carboxylate hydrochloride
tert-Butyl azetidine-2-carboxylate hydrochloride
Formel: C₈H₁₅NO₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD28975194
CAS-Nummer: 208034-97-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR