N,N-Dimethyl-2-azetidinecarboxamide hydrochloride

Lieferant: Apollo Scientific

OR451007-1G
APOSOR451007-1GEA 1160 EUR
APOSOR451007-1G
N,N-Dimethyl-2-azetidinecarboxamide hydrochloride
N,N-Dimethyl-2-azetidinecarboxamide hydrochloride
Formel: C₆H₁₂N₂O·ClH
Molecular Weight: 164,633 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28895101
CAS-Nummer: 787563-98-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR