1-(3-Methyl-3-azetidinyl)-piperidine dihydrochloride

Lieferant: Apollo Scientific

OR451008-1G
APOSOR451008-1GEA 1250 EUR
APOSOR451008-1G
1-(3-Methyl-3-azetidinyl)-piperidine dihydrochloride
1-(3-Methyl-3-azetidinyl)-piperidine dihydrochloride
Formel: C₉H₁₈N₂·2ClH
Molecular Weight: 227,175 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28975195
CAS-Nummer: 24083-68-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR