N,N,3-Trimethyl-3-azetidinamine dihydrochloride

Lieferant: Apollo Scientific

Synonyme: N,N,3-Trimethylazetidin-3-amine dihydrochloride

OR451009-1G
APOSOR451009-1GEA 1250 EUR
APOSOR451009-1G
N,N,3-Trimethyl-3-azetidinamine dihydrochloride
N,N,3-Trimethyl-3-azetidinamine dihydrochloride
Formel: C₆H₁₄N₂·2ClH
Molecular Weight: 187,111 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10565801
CAS-Nummer: 133891-76-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR