3-((Dimethylamino)methyl)azetidine dihydrochloride
Lieferant: Apollo Scientific
Synonyme:
1-(Azetidin-3-yl)-N,N-dimethylmethanamine dihydrochloride
OR451010-1G
APOSOR451010-1GEA
360
EUR
APOSOR451010-1G
3-((Dimethylamino)methyl)azetidine dihydrochloride
3-((Dimethylamino)methyl)azetidine dihydrochloride
Formel:
C₆H₁₄N₂·2ClH Molecular Weight: 187,111 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09264464 CAS-Nummer: 321890-22-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...