3-Azetidinol methanesulphonate hydrochloride

Lieferant: Apollo Scientific

OR451011-1G
APOSOR451011-1GEA 931 EUR
APOSOR451011-1G
3-Azetidinol methanesulphonate hydrochloride
3-Azetidinol methanesulphonate hydrochloride
Formel: C₄H₉NO₃S·ClH
Molecular Weight: 187,645 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08062423
CAS-Nummer: 36476-89-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR