3-(3,4-Dichlorophenoxy)azetidine hydrochloride

Lieferant: Apollo Scientific

OR451012-1G
APOSOR451012-1GEA 873 EUR
APOSOR451012-1G
3-(3,4-Dichlorophenoxy)azetidine hydrochloride
3-(3,4-Dichlorophenoxy)azetidine hydrochloride
Formel: C₉H₁₀Cl₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD09264481
CAS-Nummer: 606129-60-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR