4-(Azetidin-1-yl)piperidine dihydrochloride
Lieferant: Apollo Scientific
OR451013-1G
APOSOR451013-1GEA
1160
EUR
APOSOR451013-1G
4-(Azetidin-1-yl)piperidine dihydrochloride
4-(Azetidin-1-yl)piperidine dihydrochloride
Formel:
C₈H₁₆N₂·2ClH Molecular Weight: 213,148 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10565804 CAS-Nummer: 864246-02-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...