1-(Diphenylmethyl)-2-azetidinemethanol
Lieferant: Apollo Scientific
OR451014-1G
APOSOR451014-1GEA
1160
EUR
APOSOR451014-1G
1-(Diphenylmethyl)-2-azetidinemethanol
1-(Diphenylmethyl)-2-azetidinemethanol
Formel:
C₁₇H₁₉NO Molecular Weight: 253,339 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07786954 CAS-Nummer: 72351-68-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...