tert-Butyl azetidine-3-carboxylate hydrochloride

Lieferant: Apollo Scientific

OR451016-1G
APOSOR451016-1GEA 213 EUR
APOSOR451016-1G
tert-Butyl azetidine-3-carboxylate hydrochloride
tert-Butyl azetidine-3-carboxylate hydrochloride
Formel: C₈H₁₅NO₂·ClH
Molecular Weight: 193,671 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20528798
CAS-Nummer: 53871-08-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR