3-(Phenylmethoxy)-azetidine hydrochloride

Lieferant: Apollo Scientific

OR451018-5G
APOSOR451018-5GEA 0 EUR
APOSOR451018-5G
3-(Phenylmethoxy)-azetidine hydrochloride
3-(Phenylmethoxy)-azetidine hydrochloride
Formel: C₁₀H₁₄ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD09907646
CAS-Nummer: 897019-59-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR