3-Ethoxyazetidine hydrochloride
Lieferant: Apollo Scientific
OR451019-1G
APOSOR451019-1GEA
582
EUR
APOSOR451019-1G
3-Ethoxyazetidine hydrochloride
3-Ethoxyazetidine hydrochloride
Formel:
C₅H₁₁NO·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD06804556 CAS-Nummer: 535924-73-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...