1-Acetyl-2-azetidinecarboxylic acid

Lieferant: Apollo Scientific

OR451020-1G
APOSOR451020-1GEA 1160 EUR
APOSOR451020-1G
1-Acetyl-2-azetidinecarboxylic acid
1-Acetyl-2-azetidinecarboxylic acid
Formel: C₆H₉NO₃
Molecular Weight: 143,141 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11849023
CAS-Nummer: 474013-98-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR