1-Fmoc-3-azetidine acetic acid
Lieferant: Apollo Scientific
OR451021-1G
APOSOR451021-1GEA
1160
EUR
APOSOR451021-1G
1-Fmoc-3-azetidine acetic acid
1-Fmoc-3-azetidine acetic acid
Formel:
C₂₀H₁₉NO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD02179124 CAS-Nummer: 959236-89-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...