3-(4-Bromophenoxy)azetidine hydrochloride

Lieferant: Apollo Scientific

OR451022-1G
APOSOR451022-1GEA 960 EUR
APOSOR451022-1G
3-(4-Bromophenoxy)azetidine hydrochloride
3-(4-Bromophenoxy)azetidine hydrochloride
Formel: C₉H₁₁BrClNO
Molecular Weight: 264.56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05664840
CAS-Nummer: 1380571-76-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR