3-(4-Chlorophenoxy)azetidine hydrochloride

Lieferant: Apollo Scientific

OR451023-1G
APOSOR451023-1GEA 438 EUR
APOSOR451023-1G
3-(4-Chlorophenoxy)azetidine hydrochloride
3-(4-Chlorophenoxy)azetidine hydrochloride
Formel: C₉H₁₀ClNO·ClH
Molecular Weight: 220,096 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00277797
CAS-Nummer: 490021-97-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR