3-Cyano-3-methylazetidine hydrochloride
Lieferant: Apollo Scientific
Synonyme:
3-Methylazetidine-3-carbonitrile hydrochloride, 3-Methyl-3-azetidinecarbonitrile hydrochloride
OR451025-1G
APOSOR451025-1GEA
1080
EUR
APOSOR451025-1G
3-Cyano-3-methylazetidine hydrochloride
3-Cyano-3-methylazetidine hydrochloride
Formel:
C₅H₉ClN₂ Molecular Weight: 132,59 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11849039 CAS-Nummer: 936850-33-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...