3-Benzyl-azetidine hydrochloride

Lieferant: Apollo Scientific

Synonyme: 3-Benzylazetidine hydrochloride

OR451029-1G
APOSOR451029-1GEA 383 EUR
APOSOR451029-1G
3-Benzyl-azetidine hydrochloride
3-Benzyl-azetidine hydrochloride
Formel: C₁₀H₁₃N·ClH
Molecular Weight: 183.678 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16619075
CAS-Nummer: 7606-32-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR