3-Methyl-3-azetidinecarboxylic acid hydrochloride

Lieferant: Apollo Scientific

OR451030-1G
APOSOR451030-1GEA 1160 EUR
APOSOR451030-1G
3-Methyl-3-azetidinecarboxylic acid hydrochloride
3-Methyl-3-azetidinecarboxylic acid hydrochloride
Formel: C₅H₉NO₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD20257491
CAS-Nummer: 1365411-50-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR