1-Boc-3-amino-3-azetidinecarboxylic acid

Lieferant: Apollo Scientific

Synonyme: 3-Amino-1-[(tert-butoxy)carbonyl]azetidine-3-carboxylic acid

OR451032-1G
APOSOR451032-1GEA 1250 EUR
APOSOR451032-1G
1-Boc-3-amino-3-azetidinecarboxylic acid
1-Boc-3-amino-3-azetidinecarboxylic acid
Formel: C₉H₁₆N₂O₄
Molecular Weight: 216,234 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18375110
CAS-Nummer: 1262412-13-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR