1-Fmoc-2-azetidinecarboxylic acid

Lieferant: Apollo Scientific

OR451033-1G
APOSOR451033-1GEA 771 EUR
APOSOR451033-1G
1-Fmoc-2-azetidinecarboxylic acid
1-Fmoc-2-azetidinecarboxylic acid
Formel: C₁₉H₁₇NO₄
Molecular Weight: 323,343 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02093995
CAS-Nummer: 184763-07-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR