3-(4-Methoxyphenoxy)-azetidine hydrochloride
Lieferant: Apollo Scientific
OR451034-1G
APOSOR451034-1GEA
873
EUR
APOSOR451034-1G
3-(4-Methoxyphenoxy)-azetidine hydrochloride
3-(4-Methoxyphenoxy)-azetidine hydrochloride
Formel:
C₁₀H₁₃NO₂·ClH Molecular Weight: 215,677 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05664838 CAS-Nummer: 1236862-34-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...