tert-Butyl-3-(1-hydroxy-1-methylethyl)azetidine-1-carboxylate

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 3-(2-hydroxypropan-2-yl)azetidine-1-carboxylate, 1-Boc-3-(1-hydroxy-1-methylethyl)-azetidine

OR451036-1G
APOSOR451036-1GEA 137 EUR
APOSOR451036-1G
tert-Butyl-3-(1-hydroxy-1-methylethyl)azetidine-1-carboxylate
tert-Butyl-3-(1-hydroxy-1-methylethyl)azetidine-1-carboxylate
Formel: C₁₁H₂₁NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD22370133
CAS-Nummer: 1257293-79-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR