3-Methyl-3-phenoxyazetidine hydrochloride

Lieferant: Apollo Scientific

OR451037-1G
APOSOR451037-1GEA 1250 EUR
APOSOR451037-1G
3-Methyl-3-phenoxyazetidine hydrochloride
3-Methyl-3-phenoxyazetidine hydrochloride
Formel: C₁₀H₁₃NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD28975198
CAS-Nummer: 1323362-91-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR