1-Acetyl-3-azetidinecarboxylic acid methyl ester

Lieferant: Apollo Scientific

OR451038-5G
APOSOR451038-5GEA 1740 EUR
APOSOR451038-5G
1-Acetyl-3-azetidinecarboxylic acid methyl ester
1-Acetyl-3-azetidinecarboxylic acid methyl ester
Formel: C₇H₁₁NO₃
Molecular Weight: 157,167 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21098315
CAS-Nummer: 1353953-76-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR