1-(Methylsulphonyl)-3-azetidinecarboxylic acid

Lieferant: Apollo Scientific

OR451039-1G
APOSOR451039-1GEA 1160 EUR
APOSOR451039-1G
1-(Methylsulphonyl)-3-azetidinecarboxylic acid
1-(Methylsulphonyl)-3-azetidinecarboxylic acid
Formel: C₅H₉NO₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD16653314
CAS-Nummer: 1219828-27-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR