Methyl 1-(Methylsulphonyl)-3-azetidinecarboxylate

Lieferant: Apollo Scientific

OR451040-1G
APOSOR451040-1GEA 1160 EUR
APOSOR451040-1G
Methyl 1-(Methylsulphonyl)-3-azetidinecarboxylate
Methyl 1-(Methylsulphonyl)-3-azetidinecarboxylate
Formel: C₆H₁₁NO₄S
Molecular Weight: 193,221 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22682788
CAS-Nummer: 1418117-81-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR