N-Cyclopentyltetrahydro-2H-pyran-4-amine hydrochloride

Lieferant: Apollo Scientific

Synonyme: N-Cyclopentyl-tetrahydro-2H-pyran-4-amine

OR451046-1G
APOSOR451046-1GEA 960 EUR
APOSOR451046-1G
N-Cyclopentyltetrahydro-2H-pyran-4-amine hydrochloride
N-Cyclopentyltetrahydro-2H-pyran-4-amine hydrochloride
Formel: C₁₀H₂₀ClNO
Molecular Weight: 205.73 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11035935
CAS-Nummer: 859521-03-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR