4-[(Tetrahydro-2H-pyran-4-yl)oxy]aniline hydrochloride

Lieferant: Apollo Scientific

OR451048-5G
APOSOR451048-5GEA 1980 EUR
APOSOR451048-5G
4-[(Tetrahydro-2H-pyran-4-yl)oxy]aniline hydrochloride
4-[(Tetrahydro-2H-pyran-4-yl)oxy]aniline hydrochloride
Formel: C₁₁H₁₅NO₂·ClH
Molecular Weight: 229,703 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28975199
CAS-Nummer: 917483-71-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR