Tetrahydro-2H-thiopyran-4-methanol 1,1-dioxide

Lieferant: Apollo Scientific

OR451050-1G
APOSOR451050-1GEA 1160 EUR
APOSOR451050-1G
Tetrahydro-2H-thiopyran-4-methanol 1,1-dioxide
Tetrahydro-2H-thiopyran-4-methanol 1,1-dioxide
Formel: C₆H₁₂O₃S
Molecular Weight: 164,223 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16036707
CAS-Nummer: 473254-28-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR