1-(Tetrahydro-2H-pyran-4-yl)piperazine dihydrochloride

Lieferant: Apollo Scientific

OR451051-5G
APOSOR451051-5GEA 1900 EUR
APOSOR451051-5G
1-(Tetrahydro-2H-pyran-4-yl)piperazine dihydrochloride
1-(Tetrahydro-2H-pyran-4-yl)piperazine dihydrochloride
Formel: C₉H₂₀Cl₂N₂O
Molecular Weight: 243,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11858471
CAS-Nummer: 790223-65-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR