4-(Pyrrolidin-2-ylmethyl)morpholine
Lieferant: Apollo Scientific
OR451052-5G
APOSOR451052-5GEA
0
EUR
APOSOR451052-5G
4-(Pyrrolidin-2-ylmethyl)morpholine
4-(Pyrrolidin-2-ylmethyl)morpholine
Formel:
C₉H₁₈N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD06653314 CAS-Nummer: 215503-90-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...