(S)-4-(2-Pyrrolidinylmethyl)morpholine dihydrochloride

Lieferant: Apollo Scientific

OR451053-1G
APOSOR451053-1GEA 873 EUR
APOSOR451053-1G
(S)-4-(2-Pyrrolidinylmethyl)morpholine dihydrochloride
(S)-4-(2-Pyrrolidinylmethyl)morpholine dihydrochloride
Formel: C₉H₁₈N₂O·2ClH
Molecular Weight: 243,174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28975200
CAS-Nummer: 91790-91-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR