(S)-2-(N-Boc-aminomethyl)morpholine

Lieferant: Apollo Scientific

Synonyme: tert-Butyl N-[(2S)-morpholin-2-ylmethyl]carbamate

OR451056-500MG
APOSOR451056-500MGEA 960 EUR
APOSOR451056-500MG
(S)-2-(N-Boc-aminomethyl)morpholine
(S)-2-(N-Boc-aminomethyl)morpholine
Formel: C₁₀H₂₀N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD12545849
CAS-Nummer: 875551-59-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR